Anti-S100A9 Mouse Monoclonal Antibody [clone: 1H9]
10793-226
: Genetex
- Antibody Type:Primary
- Antigen Name:S100 Calcium Binding Protein A9
- Antigen Symbol:S100A9
- Clonality:Monoclonal
- Clone:1H9
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Size:50 μg
- Gene ID:6280
- Antigen Synonyms:CAGB|MRP14|CFAG|NIF|MAC387|MIF|L1AG|60B8AG|CGLB|P14|LIAG
- Storage Buffer:PBS, containing 0.02% sodium azide and 0.1% bovine serum albumin.
- Storage Temperature:Product should be stored at 4 °C. Under recommended storage conditions, product is stable for one year.
- Cat. No.:10793-226
Mouse Monoclonal antibody [1H9] to S100A9
Type: Primary
Antigen: S100A9
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human