Anti-PRKAA1 Rabbit Polyclonal Antibody
Catalog # 10162-926
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Protein Kinase AMP-activated Alpha 1 Catalytic Subunit
- Antigen Symbol:PRKAA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μg
- Gene ID:5562
- Antigen Synonyms:catalytic alpha-1 chain|catalytic|AMP-activated protein kinase|5'-AMP-activated protein kinase catalytic subunit alpha-1|AMPK|5'-AMP-activated protein kinase|tau-protein kinase PRKAA1|AMPK subunit alpha-1|AMPK alpha 1|AMP -activate kinase alpha 1 subunit|alpha-1|AMPKa1
- Storage Buffer:phosphate buffered saline , pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
- Storage Temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:A synthesized peptide derived from human AMPKalpha
- Purification:Antigen affinity purification
- Cat. No.:10162-926
- Supplier no.:GTX52373
Specifications
About this item
Rabbit polyclonal antibody to AMPK alpha
Type: Primary
Antigen: PRKAA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat