Anti-SLC7A9 Rabbit Polyclonal Antibody
Catalog # 10164-656
Supplier: Avantor
Specifications
- Antibody Type:Primary
- Antigen Name:Solute Carrier Family 7 Member 9
- Antigen Symbol:SLC7A9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:100 μg
- Gene ID:11136
- Antigen Synonyms:CSNU3|BAT1|FLJ94301
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and less than 0.09% sodium azide.
- Storage Temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:KLH conjugated synthetic peptide derived from human SLC7A9
- Purification:Purified by Protein A and peptide affinity chromatography.
- Cat. No.:10164-656
- Supplier no.:GTX50993
Specifications
About this item
Rabbit Polyclonal antibody to SLC7A9
Type: Primary
Antigen: SLC7A9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human