Anti-Atp5b Mouse Monoclonal Antibody [clone: 4.3E8.D10]
10168-770
: Genetex
- Antibody Type:Primary
- Antigen Name:ATP Synthase beta
- Antigen Symbol:Atp5b
- Clonality:Monoclonal
- Clone:4.3E8.D10
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μg
- Gene ID:171374
- Storage Buffer:PBS containing 0.05 % sodium azide and 1 mg/ml BSA
- Storage Temperature:–20 °C, avoid freeze/thaw cycles
- Concentration:1 mg/ml
- Immunogen:Intact rat mitochondria.
- Purification:purified
- Cat. No.:10168-770
- Supplier no.:GTX25432
Mouse monoclonal antibody [4.3E8.D10] to ATP Synthase beta
Type: Primary
Antigen: Atp5b
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat