Anti-CHRND Mouse Monoclonal Antibody [clone: 88B]
10167-988
: Genetex
- Antibody Type:Primary
- Antigen Name:AChR
- Antigen Symbol:CHRND
- Clonality:Monoclonal
- Clone:88B
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Fish,Rabbit,Amphibian,Vertebrates,Chicken,Human,Rat,Mouse
- Size:100 μl
- Gene ID:424940
- Storage Buffer:PBS containing 0.05% sodium azide.
- Storage Temperature:Keep as concentrated solution. Aliquot and store at -80ºC or below. Avoid multiple freeze-thaw cycles.
- Immunogen:Purified Torpedo californica acetylcholine receptor.
- Purification:Ascites fluid
- Cat. No.:10167-988
- Supplier no.:GTX22804
Mouse Monoclonal antibody [88B] to AChR
Type: Primary
Antigen: CHRND
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Chicken, Rat, Amphibians, Fish, Rabbit, Vertebrates