Anti-SP2 Rabbit Polyclonal Antibody
Catalog # 10166-910
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Sp2 Transcription Factor
- Antigen Symbol:SP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:6668
- Storage Buffer:1XPBS, 20% Glycerol (pH7). 0.025% ProClin 300 was added as a preservative.
- Storage Temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the center region of human SP2. The exact sequence is proprietary.
- Purification:Purified by antigen-affinity chromatography.
- Cat. No.:10166-910
- Supplier no.:GTX129030
Specifications
About this item
Rabbit Polyclonal antibody to SP2 (Sp2 transcription factor)
Type: Primary
Antigen: SP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human