Anti-WNT3 Rabbit Polyclonal Antibody
Catalog # 10167-090
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Wingless-Type Mmtv Integration Site Family, Member 3
- Antigen Symbol:WNT3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:7473
- Antigen Synonyms:TETAMS|INT4
- Storage Buffer:1XPBS, 20% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
- Storage Temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the center region of human WNT3. The exact sequence is proprietary.
- Purification:Purified by antigen-affinity chromatography.
- Cat. No.:10167-090
- Supplier no.:GTX128100
Specifications
About this item
Rabbit Polyclonal antibody to WNT3 (wingless-type MMTV integration site family, member 3)
Type: Primary
Antigen: WNT3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human