Order Entry
United States
ContactUsLinkComponent
 
Anti-NOXA1 Rabbit Polyclonal Antibody
Catalog # 10165-390
Supplier:  Genetex
Anti-NOXA1 Rabbit Polyclonal Antibody
Catalog # 10165-390
Supplier:  Genetex
Supplier Number:  GTX12206

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Nadph Oxidase Activator 1
  • Antigen Symbol:
    NOXA1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 μg
  • Gene ID:
    10811
  • Antigen Synonyms:
    NY-CO-31|SDCCAG31|p51NOX
  • Storage Buffer:
    5% BSA, 0.9% NaCl, 0.2% Na2HPO4, 0.05% Thimerosal, 0.05% NaN3.
  • Storage Temperature:
    Keep as concentrated solution. Aliquot and store at –20 °C . Avoid multiple freeze-thaw cycles.
  • Concentration:
    0.50 mg/ml
  • Immunogen:
    A synthetic peptide corresponding to a sequence in the middle region of human NOXA1.
  • Purification:
    Immunogen affinity purified.
  • Cat. No.:
    10165-390
  • Supplier no.:
    GTX12206

Specifications

About this item

Rabbit Polyclonal antibody to NOXA1

Type: Primary
Antigen: NOXA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human