Anti-GABRA1 Mouse Monoclonal Antibody [clone: S95-35]
Catalog # 10169-710
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:GA binding Protein transcription Factor Alpha Subunit 1
- Antigen Symbol:GABRA1
- Clonality:Monoclonal
- Clone:S95-35
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human,Rat,Mouse
- Size:100 μg
- Antigen Synonyms:GABA(A) receptor subunit alpha-1|Gamma-aminobutyric acid receptor subunit alpha-1|GABRA1|ECA4|EIEE19|EJM5|EJM
- Storage Buffer:PBS pH7.4, 50% glycerol, 0.09% sodium azide
- Storage Temperature:Store at -20C or below.
- Concentration:1 mg/ml
- Immunogen:Fusion protein amino acids 355-394 of mouse GABA-A-R-Alpha1, accession number P62812
- Purification:Protein G purified
- Cat. No.:10169-710
- Supplier no.:GTX10698
Specifications
About this item
Mouse monoclonal antibody [S95-35] to GABA(A) Receptor Alpha1
Type: Primary
Antigen: GABRA1
Clonality: Monoclonal
Clone: S95-35
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2A
Reactivity: Human, Mouse, Rat