Specifications
- Target protein:Gluten
- Description:ALLER-TEK™ Gluten ELISA Kits, Elisa Technologies Inc
- Size:96 microwell plate system with single strip format (12 strips of 8 wells)
- Time to Results:40 minutes for preparation/extraction; 130 minutes for immunoassay
- Shelf Life:1 year from date of manufacture
- Detection Limit:5 ppm
- Cat. No.:102963-546
- Supplier no.:510821
- No. of tests:Up to 40 samples in duplicate
Specifications
About this item
The ALLER-TEK™ Gluten ELISA was designed to quantitate low levels of total gluten in food ingredients as well as in prepared and processed foods and beverages.
- Standard curve for wheat gluten for quantitation
- Recognizes both the gliadin and glutenin fractions of gluten
- Accurate determination of total gluten levels in food
The ALLER-TEK™ Gluten ELISA was designed to quantitate low levels of total gluten in food ingredients as well as in prepared and processed foods and beverages. By using a specific antibody, this assay recognizes both the gliadin and glutenin fractions of gluten to provide an accurate determination of total gluten levels in food and food products. The kit includes a standard curve for wheat gluten for quantitation between 5 and 80 ppm. For the accurate quantitation of barley gluten in samples suspected to contain barley but not wheat, rye or other related grains, a separate set of barley standards is available.