ContactUsLinkComponent
ELISA-TEK™ Cooked Meat Speciation Kits Elisa Technologies Inc
ELISA-TEK™ Cooked Meat Speciation Kits Elisa Technologies Inc
Certificates
ELISA-TEK™ Cooked Meat Speciation Kits Elisa Technologies Inc
Specifications
  • Time to Results:
    ~ 30 minutes (for sample preparation/extraction) 3 hours for immunoassay procedures
  • Shelf Life:
    Up to 12 months from date of manufacture
  • Detection Limit:
    1.0% in canned, cooked, or processed foods (USDA protocol)