Ward's® Live GFP producing E. coli Culture
Catalog # 470179-088
Supplier: Avantor
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Description:E. coli Strain B21
- Storage conditions:Refrigerator
- Cat. No.:470179-088
- Packaging:Plate
- Media:Luria agar with ampicillin
- Supplier no.:WARD470179-088
- Incubation conditions:37 °C
Specifications
About this item
Eschericia coli culture that produces GFP, visible in the presence of UV light.
Ordering information: These products contain living or perishable materials and will ship via overnight or second day delivery. Please specify a delivery date during checkout
Bacterial and fungal cultures are restricted and can only be ordered by educational professionals.
Bacterial and fungal cultures are restricted and can only be ordered by educational professionals.
Caution: Although this organism is non pathogenic, Ward’s recommends that you utilize proper aseptic technique when handling all microorganisms.