Order Entry
United States
ContactUsLinkComponent
Consumables for SafTest Platforms, MP Biomedicals
Consumables for SafTest Platforms, MP Biomedicals
Supplier:  MP Biomedicals
Certificates
undefined
Consumables for SafTest Platforms, MP Biomedicals
Supplier:  MP Biomedicals
Certificates

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Storage Temperature:
    RT
  • Application:
    Oil/Fat Degradation
  • Storage Conditions:
    RT
  • Instrument Compatibility:
    SafTest Vacuum Pump
  • Application Areas:
    Food Quality
  • Lab Essential Type:
    Vacuum Filtration
  • Test Parameters:
    AciSafe MSA; AldeSafe MSA; AldeSafe STD; AlkalSafe STD; FaSafe HSY; FaSafe STD; Percent Fat; PeroxySafe HSY; PeroxySafe MSA; PeroxySafe STD; Total Glycerin
  • Product Type:
    Lab Essentials

Specifications

Related Information

Frequently Bought Together