Order Entry
United States
ContactUsLinkComponent
Nitric acid 69%, Veritas®, redistilled
Oxidizing
Corrosive
Health
Nitric acid 69%, Veritas®, redistilled
Supplier:  GFS Chemicals
Certificates
Oxidizing
Corrosive
Health
Nitric acid 69%, Veritas®, redistilled
Supplier:  GFS Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 68.0%
  • Chloride:
    0.00001%
  • Sulfate:
    0.00001%
  • Arsenic:
    0.0000001%
  • Heavy Metals:
    0.000001%
  • Iron:
    0.000001%
  • Residue after Ignition:
    0.0001%
  • Color:
    Max. 10 APHA

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together