STAT-MAT® Absorbent, PIG®
56620-542
: New Pig
- Description:PIG® STAT-MAT® Absorbent
- Cat. No.:56620-542
- Absorbency:40.2 gal./roll
- Supplier no.:MAT212
Heavyweight for increased durability and absorbency.
- Dissipates static buildup
- Deters static charge during transportation
- Increased durability and absorbency
Heavyweight for increased durability and absorbency. Treated with a topical solution to dissipate static buildup. Packaged in special static-dissipative bags to deter a static charge during transportation.