Order Entry
United States
Orders LinkContactUsLinkComponent
API® Suspension Medium, Biomerieux

API® Suspension Medium, Biomerieux

API® Suspension Medium, 100×2 ml, Ampoule
Catalog # 95060-980
Supplier:   bioMerieux
Supplier #:  70700
API® Suspension Medium, Biomerieux
Catalog # 95060-980
Supplier:  bioMerieux
CAS Number:  
API® Suspension Medium, Biomerieux

API® Suspension Medium, 100×2 ml, Ampoule

CAS Number:  
Price
$319.42
Log in for availability
Pack of 2
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Reagents for use with various API® kits.

  • Economical
  • Easy-to-use
  • For use with various kits

This economical suspension medium comes in 100 x 2 mL ampules. Medium can be used with a number of other API® kits.

Documents

Specifications

Specifications

  • Pack type:
    Ampoule
  • Description:
    API® Suspension Medium
  • Size:
    100×2 ml

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.