Smart USB UPS System, Tripp Lite
Supplier: Tripp Lite
Certificates
About this item
Intelligent, line-interactive battery backup for workstations.
- Supports battery replacement via built-in access panel
- Attractive stealth black tower configuration
- 3 year manufacturers warranty
- Built-in audible alarm and 4 front panel LEDs indicate UPS system and line power status
Tripp Lite's SMARTPro line interactive UPS system offers voltage regulation, surge suppression and long lasting battery support for personal computers, internetworking equipment and other sensitive electronics. Internal UPS circuits support entry level PC systems for up to 30 minutes during power failures so that connected equipment can be shutdown without data loss. Automatic Voltage Regulation (AVR) circuits regulate brownouts as low as 89 volts back to usable levels without using battery power.
Certifications: Tested to UL1778 (USA); Tested to CSA (Canada); Tested to NOM (Mexico); Meets FCC Part 15 Category B (EMI); Meets FCC Part 68 / Industries Canada (telecommunications); ROHS (Reduction of Hazardous Substances).
Delivery: Package includes SMARTPro UPS System, USB and Telephone interconnection cables, Instruction manual with warranty information.
Specifications
- Catalog No:
- 10144-242
- 10144-246
- Operating Temperature:
- 0 to 40° C (32 to 104°F)
- 0 to 40° C (32 to 104°F)
- Weight:
- 14.4 lbs.
- 20 lbs.