ContactUsLinkComponent
Inlet Manifold Kit for Waters HPLC Systems, Restek
Inlet Manifold Kit for Waters HPLC Systems, Restek
Catalog # 10853-710
Supplier:  Avantor
undefined
Inlet Manifold Kit for Waters HPLC Systems, Restek
Catalog # 10853-710
Supplier:  Avantor
Specifications
  • Model:
    M45, 501, 510, 515, 590, 600, 610
  • Description:
    Inlet Manifold Kit for Waters M45, 501,510, 590, 600(E), Similar to Waters #WAT060448
  • Cat. No.:
    10853-710
  • Includes:
    TFE ball plug, TFE seat, ruby ball, manifold insert, TFE insert seal, Belleville washer (2), flat washer, inlet tube body assembly
  • Supplier no.:
    25412