ContactUsLinkComponent
Survival Kit for HPLC, Stainless Steel, Restek
Survival Kit for HPLC, Stainless Steel, Restek
Catalog # 10853-628
Supplier:  Restek
undefined
Survival Kit for HPLC, Stainless Steel, Restek
Catalog # 10853-628
Supplier:  Restek
Supplier Number:  25097
Specifications
  • Description:
    Stainless Steel Survival Kit for HPLC
  • Material:
    Stainless steel
  • Cat. No.:
    10853-628
  • Supplier no.:
    25097
Specifications
About this item
The stainless steel survival kit contains a wide range of tubing, fittings, and tools necessary to set up and maintain your HPLC system.
Kit includes a selection of lengths and IDs of 1/16" tubing, plus nuts, ferrules, a ValvTool wrench, and a zero-dead-volume union.
Ordering information: Kit includes HPLC Capillary Tubing, SS 1/16"×0.005"×5 cm, 3-pk; HPLC Capillary Tubing, SS, 1/16"×0.005"×10 cm, 3-pk; HPLC Capillary Tubing, SS, 1/16"×0.005"×20 cm, 3-pk; HPLC Capillary Tubing, SS, 1/16"×0.005"×30 cm, 3-pk; HPLC Capillary Tubing, SS, 1/16"×0.007"×5 cm, 3-pk; HPLC Capillary Tubing, SS, 1/16"×0.007"×10 cm, 3-pk; HPLC Capillary Tubing, SS, 1/16"×0.007"×20 cm, 3-pk; HPLC Capillary Tubing, SS, 1/16"×0.007"×30 cm, 3-pk; HPLC Capillary Tubing, SS, 1/16"×0.010"×5 cm, 3-pk; HPLC Capillary Tubing, SS, 1/16"×0.010"×10 cm, 3-pk; HPLC Capillary Tubing, SS, 1/16"×0.010"×20 cm, 3-pk; HPLC Capillary Tubing, SS, 1/16"×0.010"×30 cm, 3-pk; HPLC Capillary Tubing, SS, 1/16"×0.020"×5 cm, 3-pk; HPLC Capillary Tubing, SS, 1/16"×0.020"×10 cm, 3-pk; HPLC Capillary Tubing, SS, 1/16"×0.020"×20 cm, 3-pk; HPLC Capillary Tubing, SS, 1/16"×0.020"×30 cm, 3-pk; 1/16" Rheodyne Style Nut, 10-pk; 1/16" Rheodyne Style Ferrule, 10-pk; ValvTool Wrench, ea; Ferrules, 1/16" Stainless Steel, 10-pk; Nuts, 1/16" Stainless Steel, 10-pk; Zero-Dead-Volume Internal Union, ea.