Microliter™ 700 Series Syringes with Luer Tip, Stoelting
Supplier: Stoelting
Certificates
About this item
The 700 series syringe is the original hand-fitted Hamilton syringe.
This series of syringes is ideal for dispensing volumes from 0.5 μL up to 500 μL.
Ordering information: A variety of different terminations and needle options are available in this series.
Frequently Bought Together