Blue Light Conversion Screen
Blue light conversion screen, Azure
Catalog # 75794-848
Supplier: Azure Biosystems
Supplier #: AC1025
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Blue light conversion screen for transillumination of blue light.
- Transillumination of blue light
- Converts UV to blue light
- Used for imaging in gel documentation systems
Blue light conversion screen for transillumination of blue light. Converts UV to blue light for imaging in gel documentation systems.
Documents
Azure Biosystems™ Product Catalog
Product Literature
Specifications
- Description:Blue light conversion screen, Azure