Order Entry
United States
ContactUsLinkComponent
Autosampler Vials, PFA, PerkinElmer
Autosampler Vials, PFA, PerkinElmer
Catalog # 10139-362
Supplier:  PerkinElmer
undefined
Autosampler Vials, PFA, PerkinElmer
Catalog # 10139-362
Supplier:  PerkinElmer
Supplier Number:  N0777401

Specifications

  • Cap included:
    No
  • Capacity:
    4.00 ml
  • Description:
    Autosampler Vial, Graduated
  • Material:
    PFA
  • Cat. No.:
    10139-362
  • Diameter ×H:
    14×60 mm
  • Supplier no.:
    N0777401

Specifications

About this item

PerkinElmer produces a wide range of PFA autosampler vials in a variety of shapes and sizes, to cover virtually every application.

  • Preferred for semiconductor use
  • Translucent siding for optimal visibility
  • Conical interior

PFA vials are preferred for use with semiconductors, and PFA alternatives exist for a majority of polypropylene vials.