J.T.Baker® BAKER INSTRA-ANALYZED®, AA Standards, Palladium
Catalog # JT6462-4
Supplier: Avantor
CAS Number:
MOC: Non-RegulatedCertificates
Specifications
- Concentration:1000 µg/ml
- Matrix:20% HCl
- Pack type:Polyethylene bottle
- Size:150 ml
- Cat. No.:JT6462-4
- Element:Palladium
- Supplier no.:6462-04
Specifications
About this item
The standard is prepared by the careful dilution of the metal or its salt, usually 99.99% pure as verified by arc emission spectrometry.
- Packaged at a concentration of 1000 µg/ml
- Standardised with NIST standard chemicals
- Maximum shelf life and stability
The final concentration of each lot is determined by titrimetry or gravimetry.