ContactUsLinkComponent
HRP Stripping Buffer, Azure Biosystems
HRP Stripping Buffer, Azure Biosystems
Catalog # 75794-868
Supplier:  Azure Biosystems
Product available on GSA Advantage®
CAS Number:  
undefined
HRP Stripping Buffer, Azure Biosystems
Catalog # 75794-868
Supplier:  Azure Biosystems
Supplier Number:  AC2154
Product available on GSA Advantage®
CAS Number:  
Specifications
  • Description:
    Stripping Buffer for HRP Western blots
  • Size:
    500 ml
  • Cat. No.:
    75794-868
  • Supplier no.:
    AC2154
Specifications
About this item
HRP Stripping Buffer, Uniformly removes antibodies from developed HRP Western blots so the blot can be reprobed with another set of antibodies