Silver Standard for ICP, Certipur®, MilliporeSigma
EM1.70352.0100
:
- Concentration:1000 µg/ml
- Matrix:2-3 % HNO₃
- Pack type:Plastic Bottle
- Size:100 ml
- Hazard Class Description:Corrosive Material
- Cat. No.:EM1.70352.0100
- pH:0.5 (H₂O, 20°C)
- Element:Silver
- Supplier no.:1.70352.0100
Traceable to SRM from NIST.
: The quality control of ICP single-element standards is carried out by the accredited calibration laboratory according to ISO 17025.
: Includes a Certificate of Analysis, with exact data on content including uncertainty budget, trace element impurities, composition, traceability, date of release and minimum shelf life.