Order Entry
United States
ContactUsLinkComponent
Niobium Standard for ICP, Certipur®, MilliporeSigma
Niobium Standard for ICP, Certipur®, MilliporeSigma
Catalog # EM1.70337.0100
Supplier:  MilliporeSigma
CAS Number:  
Niobium Standard for ICP, Certipur®, MilliporeSigma
Catalog # EM1.70337.0100
Supplier:  MilliporeSigma
Supplier Number:  1.70337.0100
CAS Number:  

Specifications

  • Concentration:
    1000 µg/ml
  • Matrix:
    2-3 % HNO₃
  • Pack type:
    Plastic Bottle
  • Size:
    100 ml
  • Hazard Class Description:
    Corrosive Material
  • Cat. No.:
    EM1.70337.0100
  • pH:
    3 (H₂O, 20°C)
  • Element:
    Niobium
  • Supplier no.:
    1.70337.0100

Specifications

About this item

NH₄NbF₆ in H₂O

Traceable to SRM from NIST.

Certifications: The quality control of ICP single-element standards is carried out by the accredited calibration laboratory according to ISO 17025.
Ordering information: Includes a Certificate of Analysis, with exact data on content including uncertainty budget, trace element impurities, composition, traceability, date of release and minimum shelf life.