About this item
Affinity Chromatography (AFC) offers the greatest potential specificity and selectivity for the isolation or purification of biomolecules. Almost all biological molecules can be purified on the basis of a specific interaction between their chemical or biological structure and a suitable affinity ligand.
- The choice of a specific ligand is dictated by the expected interaction between the sample and the bonded phase of the resin used
- The TSKgel affinity column line consists of two group specific ligands and one chemically active functionality
Group Specific Ligands: TSKgel® Boronate-5PW, applied for carbohydrates, nucleic acids, nucleosides, nucleotides, catecholamines. TSKgel® Chelate-5PW, applied for immunoglobulins, transferrin, lectins, milk proteins, membrane proteins, peptides.
Chemically Activated Ligands: TSKgel® Tresyl-5PW for coupling of ligands to form a custom affinity resin.