Order Entry
United States
ContactUsLinkComponent
Natural Gas Standard #2, Restek
Natural Gas Standard #2, Restek
Catalog # 75795-214
Supplier:  Restek
CAS Number:  
Natural Gas Standard #2, Restek
Catalog # 75795-214
Supplier:  Restek
Supplier Number:  34439
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Concentration:
    Mol
  • Description:
    Natural Gas Standard #2
  • Size:
    13.16 L
  • Shelf Life:
    One year
  • Pressure:
    200 psig
  • Cat. No.:
    75795-214
  • Accuracy:
    Analytical accuracy: ±2%
  • Tolerance:
    Blend tolerance: ±2%
  • Supplier no.:
    34439

Specifications

About this item

Natural gas standards with 10 components are available in three mixes, from lean to rich.

  • Cylinder: 13.16 L at 200 psig
  • Mini-regulator designed specially for these standards is also available

Each of these natural gas standards has an extended list of C6+ components.

Ordering information: Natural Gas Standard #2 contains the following compounds: Nitrogen, 2.500 %; Carbon dioxide, 1.000 %; Methane UHP, 85.250 %; Ethane UHP, 5.000 %; Propane, 3.000 %; Isobutane, 1.000 %; N-Butane, 1.000 %; Isopentane, 0.500 %; N-Pentane, 0.500 %; Hexanes plus**, 0.250 %.

Available in three mixes, Standards #1 to #3, from lean to rich. Each has an extended list of C6+ components..
Packaging: Ideal Heating Value: Dry BTU/SCF @ 14.696psia & 60°F.