Potassium sulfate, Supelco®
EM1.05153.9025
:
- Pack type:Fibre carton
- Size:25 kg
- Cat. No.:EM1.05153.9025
- Supplier no.:1.05153.9025
:
:174.26 g/mol
:1698 °C (1013 hPa)
:1069 °C
:2.66 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ300
:7778-80-5
:K₂SO₄
:MFCD00011388