Order Entry
United States
ContactUsLinkComponent
tri-Sodium citrate dihydrate 99.0-101.0% (by perchloric acid titration) [99.0-101.0% anhydrous basis (perchloric acid titration)], Supelco®
tri-Sodium citrate dihydrate 99.0-101.0% (by perchloric acid titration) [99.0-101.0% anhydrous basis (perchloric acid titration)], Supelco®
Catalog # EM1.06448.9025
Supplier:  MilliporeSigma
CAS Number:  
undefined
tri-Sodium citrate dihydrate 99.0-101.0% (by perchloric acid titration) [99.0-101.0% anhydrous basis (perchloric acid titration)], Supelco®
Catalog # EM1.06448.9025
Supplier:  MilliporeSigma
Supplier Number:  1.06448.9025
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Fibre carton
  • Size:
    1 kg
  • Cat. No.:
    EM1.06448.9025
  • Supplier no.:
    1.06448.9025

Specifications

About this item

 :
 :294.1 g/mol
Melting Pt:>300 °C (lit.)
Density:1.76 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ300
Storage Temperature:Ambient
CAS Number:6132-04-3
Formula:Na₃C₆H₅O₇·2H₂O
MDL Number:MFCD00150031