Order Entry
United States
ContactUsLinkComponent
G418 disulfate 50 mg/mL in aqueous solution
G418 disulfate 50 mg/mL in aqueous solution
Catalog # 75794-854
Supplier:  IBI Scientific
CAS Number:  
undefined
G418 disulfate 50 mg/mL in aqueous solution
Catalog # 75794-854
Supplier:  IBI Scientific
Supplier Number:  IB02060
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    20 ml
  • Environmentally Preferable:
  • Cat. No.:
    75794-854
  • Supplier no.:
    IB02060

Specifications

Specification Test Results

  • Appearance:
    Clear, colorless liquid
  • Sterile filtration:
    No microbial growth

Specification Test Results

About this item

Synonyms: G418 disulfate, G-418 sulfate, G-418 sulphate, Geneticin®, G-418 disulphate, G418 sulphate, G-418 disulfate
 :
 :692.72 g/mol
EINECS:213-791-2
 :GHS08
Storage Temperature:Freezer
CAS Number:108321-42-2
Formula:C₂₀H₄₄N₄O₁₈S₂
MDL Number:MFCD00058314