Syringe Filters, Nylon, Bonna-Agela Technologies
Supplier: Phenomenex
Certificates
About this item
These Nylon membrane filters feature housing made of high purity polypropylene.
- Autoclavable
- Syringe filters are designed for use in both directions
- Female Luer-Lock inlets secure the fit to syringes
- Good flexibility and chemical compatibility
Frequently Bought Together