ContactUsLinkComponent
Gas Anesthesia Platform for Mice, Stoelting
Gas Anesthesia Platform for Mice, Stoelting
Catalog # 100229-056
Supplier:  Avantor
Minority or Woman-Owned Business Enterprise
Gas Anesthesia Platform for Mice, Stoelting
Catalog # 100229-056
Supplier:  Avantor
Minority or Woman-Owned Business Enterprise
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Gas anesthesia platform and mouse gas mask adaptor
  • Cat. No.:
    100229-056
  • Supplier no.:
    50264
Specifications
About this item
This anesthesia platform provides precise and secure positioning of an animal during anesthesia and surgery.
  • Automatic and effortless positioning of animal
After removing the animal from the induction chamber, it can be quickly mounted on the platform. The animal’s teeth are placed over the incisor bar and the mouse mask slides forward until the cone is snug around the animal’s nose, thereby substituting for the nose clamp. The gas anesthesia platform and mask is also useful for non-stereotaxic related experimental applications. With the platform placed on the bench top, the head of the mouse or rat can be fixed in the mask, appropriate tubing (¼" ID) can be attched to the gas mask, and it's ready to perform surgery, injections, or implantations under gas anesthesia.