Coverage® Spray HB Plus Disinfectant, STERIS®
14212-963
: Avantor
- Description:Coverage® Spray HB Plus Disinfectant
- Size:650 mL (22 fl. oz.)
- Cat. No.:14212-963
- Supplier no.:162477
Coverage Spray HB is a ready-to-use one-step disinfectant that has a broad range of activity against Gram-negative and Gram-positive bacteria, viruses including Hepatitis B (HBV), Hepatitis C (HCV), HIV-1, and fungi.
- Hospital-grade quat that kills Hepatitis B Virus in one step
- Kills HIV in 30 seconds
- Clean, fresh lemon fragrance
- Convenient, easy-to-hold trigger sprayer
- Cost-effective, no mixing, no dilution errors
- EPA-registered
Coverage Spray HB Ready-to-Use Disinfectant Cleaner is recommended for use on toilet seats, telephones, doorknobs, light switches, furniture, and other hard nonporous surfaces such as walls and floors. It is intended for use in hospitals (including laboratories, urgent care facilities, doctors offices, and dialysis centers), day care centers, and veterinary clinics.