ContactUsLinkComponent
Vacuum Filtration Adapters, with PTFE Stopcock, Chemglass
Vacuum Filtration Adapters, with PTFE Stopcock, Chemglass
Catalog # 33502-686
Supplier:  Chemglass
Small Business Enterprise
undefined
Vacuum Filtration Adapters, with PTFE Stopcock, Chemglass
Catalog # 33502-686
Supplier:  Chemglass
Supplier Number:  CG-1053-A-02
Small Business Enterprise
Specifications
  • [ST] joint:
    24/40
  • Description:
    Vacuum Filtration Adapter, accepts Buchner Funnel
  • Cat. No.:
    33502-686
  • Pluro stopper:
    2
  • Fits:
    60 ml funnel capacity
  • Supplier no.:
    CG-1053-A-02
Specifications
About this item
Adapter is used for reduced pressure filtrations using plain stem buchner funnels.
  • With stopcock
  • Top of adapter is tooled to accept pluro-stopper
The stopcock permits control of vacuum source. The top of the adapter is tooled to accept the listed size of pluro-stopper.
Ordering information: Adapters are not supplied with rubber pluro stoppers.