Pressure Gauge Needle for Vials, PerkinElmer
Catalog # 10132-532
Supplier: PerkinElmer
Specifications
- Description:Pressure Gauge with Needle, for Vials
- Cat. No.:10132-532
- Supplier no.:B0501377
Specifications
About this item
For convenient tightening and loosening of the screw connectors used with PTFE tubing assemblies.
- Easy to use
- Durable
- Reliable
Pressure gauge needle is designed to measure air pressure in vials.