Radleys Carousel 12 Plus™ Reaction Station Systems, Heidolph
Supplier: Heidolph NA, LLC
Certificates
About this item
The unique patented Carousel 12 Plus™ simultaneously heats or cools, stirs, and refluxes multiple samples under an inert atmosphere
- Accepts up to 12 glass tubes with a reaction volume from 1 mL to 20 mL
- Powerful, even stirring
- Fits onto a Carousel stirring hotplate
- Rapid heating to 220 °C and cooling to –78 °C
- Compact size offers small benchtop footprint for easy storage
- Visibility slots allow easier viewing of tube contents during experiments
- Fluoropolymer coating for chemical resistance and easy cleaning
Removable Reflux head allows reaction tubes to be transferred between heated base, cooled base or stand without disconnecting the tubes. Chemically resistant PFA caps feature a quick-thread for fast attachment to the glass tubes and easy-on push connections to the stainless steel gas outlets. Unique removable PFA insulation plate helps insulate the heated base for faster heating and energy savings of up to 36%.
It is an effective personal synthesis station for parallel solution phase chemistry and solid supported reagent based synthesis.
Ordering information: The Basic System includes Carousel 12 Plus™ Reaction Station, Quick-Thread Glass Reaction pk 6, Quick-Thread Glass Reaction Tube, pk 12, (2) Easy-On Fluoropolymer Caps, pk 6, Octagonal Stirring Bar 13 mm, pk 20, Cross Shape Stirring Bar 16.5 mm RE, pk 20, PTFE Magnetic Stirring Bar Retriever 350 mm, PTFE Magnetic Stirring Bar Evaluation Kit, pk 30, Silicone Suba Seals for All Caps, pk 100, Zinc Plated Wire Rack for 36 x Tubes. The Standard System includes all components of the Basic System plus Carousel 12 Plus™ Stand, MR Hei-Tec Magnetic Stirrer, and Pt 1000 Temperature Sensor.
Frequently Bought Together