Order Entry
United States
ContactUsLinkComponent
Adipocyte FABP, AFABP, (FABP4), Human ELISA, BioVendor
Adipocyte FABP, AFABP, (FABP4), Human ELISA, BioVendor
Catalog # 101215-168
Supplier:  BioVendor
CAS Number:  
Adipocyte FABP, AFABP, (FABP4), Human ELISA, BioVendor
Catalog # 101215-168
Supplier:  BioVendor
Supplier Number:  RD191036200R
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Target protein:
    Adipocyte-type fatty acid-binding protein, A-FABP, Fatty acid-binding protein 4, Adipocyte lipid-binding protein, ALBP, FABP4
  • Description:
    Adipocyte FABP (FABP4) human ELISA
  • Size:
    1 kit
  • Sample Type:
    Serum, plasma-EDTA, plasma-heparin, plasma-citrate
  • Detection Method:
    Colorimetric
  • Shelf Life:
    See expiration date
  • Detection Range:
    0.5 - 25 ng/ml
  • Storage Temperature:
    2…8 °C
  • Sample Volume:
    20 µl
  • Sensitivity:
    0.05 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    101215-168
  • Supplier no.:
    RD191036200

Specifications

About this item

96 wells (1 kit) - ELISA is a competitive enzyme immunoassay for the quantitative measurement of AFABP4.

  • Calibration Range: 0.5-25 ng/ml
  • Limit of Detection: 0.05 ng/ml
  • Plasma-citrate, plasma-EDTA, plasma-heparin, aerum: 20 µl/well

Intended use -The RD191036200R Human Adipocyte FABP ELISA is a sandwich enzyme immunoassay for the quantitative measurement of human AFABP. Adipocyte fatty acid binding protein AFABP is a 15 kDa member of the intracellular fatty acid binding protein (FABP) family, which is known for the ability to bind fatty acids and related compounds (bile acids or retinoids) in an internal cavity. AFABP is expressed in a differentiationdependent fashion in adipocytes and is a critical gene in the regulation of the biological function of these cells. In the BioVendor Human AFABP ELISA, standards, quality controls and samples are incubated in microplate wells pre-coated with polyclonal anti-human AFABP antibody. After 60 minutes incubation and washing, biotin labelled polyclonal anti-human AFABP antibody is added and incubated for 60 minutes with captured AFABP. After another washing, streptavidin-HRP conjugate is added. After 30 minutes incubation and the last washing step, the remaining conjugate is allowed to react with the substrate solution (TMB). The reaction is stopped by addition of acidic solution and absorbance of the resulting yellow product is measured. The absorbance is proportional to the concentration of AFABP. A standard curve is constructed by plotting absorbance values against concentrations of standards, and concentrations of unknown samples are determined using this standard curve.

Caution: This is intended for research use only.