Kinetic Theory and Specific Heat of Gases
Catalog # 470220-092
Supplier: Klinger
Specifications
- Description:Kinetic Theory and Specific Heat of Gases
- Cat. No.:470220-092
- Supplier no.:P2531
Specifications
About this item
Students determine the adiabatic coefficient of air.
- Oscillation Tube Included
- Stopwatch Included
- Aneroid Barometer Included
- Plastic Beaker Included
- Students Investigate the Kinetic Theory of a Gas
The experiment determines the adiabatic exponent of air from the oscillation period of a ball that caps and seals a gas volume in a glass tube. The oscillation of the ball around the equilibrium position causes adiabatic changes in the state of the gas. In the equilibrium position, the force of gravity and the opposing force resulting from the pressure of the enclosed gas are equal.
A deflection from the equilibrium position by Δx causes the pressure to change by ∆p, the cross section of the riser tube, which returns the ball to the equilibrium position.
Ordering information: This kit includes one oscillation tube with a Marriott's bottle, mechanical and hand-held stop watch, aneroid barometer , 1000 mL plastic beaker, and 50 g of Vaseline.