Order Entry
United States
ContactUsLinkComponent
 
Monocyte Chemotactic Protein-1, MCP-1, Mouse ELISA, BioVendor
Catalog # 102552-480
Supplier:  BioVendor
CAS Number:  
undefined
Monocyte Chemotactic Protein-1, MCP-1, Mouse ELISA, BioVendor
Catalog # 102552-480
Supplier:  BioVendor
Supplier Number:  RAF080R
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Primary antibody reactivity:
    Mouse
  • Target protein:
    MCP-1 (CCL2)
  • Description:
    Monocyte Chemotactic Protein-1 (MCP-1) Mouse ELISA
  • Size:
    1 kit
  • Regulatory Status:
    RUO
  • Cat. No.:
    102552-480
  • Supplier no.:
    RAF080R

Specifications

About this item

Monocyte chemotactic protein-1 (MCP-1) is a member of the C-C family of chemokines.

  • Calibration Range: 15.6-1000 pg/mL
  • Limit of Detection: 2.2 pg/mL
  • Cell culture supernatant, Serum: 50 uL/well

Monocyte chemotactic protein-1 (MCP-1) is a member of the C-C family of chemokines. Chemokines are mediators that share the capacity to induce directional migration of leukocytes. MCP-1 is a chemoattractant for many cell types such as monocytes, neutrophils, hepatic stellate cells, granulocytes and T lymphocytes. MCP-1 is believed to play a role in a variety of pathological conditions, including inflammatory and immune reactions, atherosclerosis and neoplasia. Monocyte Chemotactic Protein-1 (MCP-1) is also known as Monocyte Chemotactic Activating Factor (MCAF) or CCL2.

Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.