ContactUsLinkComponent
PURELAB® Conditioning Cartridge, ELGA LabWater
PURELAB® Conditioning Cartridge, ELGA LabWater
Catalog # 89221-822
Supplier:  ELGA LabWater
Product available on GSA Advantage®
undefined
PURELAB® Conditioning Cartridge, ELGA LabWater
Catalog # 89221-822
Supplier:  ELGA LabWater
Supplier Number:  LC178
Product available on GSA Advantage®
Specifications
  • Description:
    Conditioning Cartridge
  • Cat. No.:
    89221-822
  • Supplier no.:
    LC178
Specifications
About this item
The conditioning cartridge is for use in PURELAB® Option-E 5/10 water purification systems, for the conditioning stage required for reversed-phase sorbents.
  • Ensures ongoing water purification effectiveness
  • Provides essential conditioning of circulating system
The conditioning cartridge is easy to install and convenient to replace.