- Source:E. coli
- Size:10 mg
- Enzyme Name:beta-Lactamase
- Cat. No.:101424-064
- Supplier no.:RP179351
Beta-lactamase is a type of enzyme (EC 3.5.2.6) produced by some bacteria that is responsible for their resistance to beta-lactam antibiotics like penicillins, cephalosporins, cephamycins and carbapenems
These antibiotics have a common element in their molecular structure: a four-atom ring known as a beta-lactam. The lactamase enzyme breaks that ring open, deactivating the molecule''s antibacterial properties.