Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Source:E. coli
- Species:Human
- Size:10 µg
- Protein Synonyms:Type I interferon|MGC96956|IFNB|IFB|B cell interferon|Leukocyte interferon|IFF|IFNB1|IFN-b 1b
- Protein/Peptide Name:Rhu Interferon-Beta 1B
- Cat. No.:101423-126
- Supplier no.:RP1762340010
Interferon beta 1b Human Recombinant produced in E.Coli is a single, non-glycosylated mutein (variant form) of human Interferon beta-1b polypeptide chain containing 165 amino acids and having a molecular mass of 18510.86 Dalton.