Order Entry
United States
ContactUsLinkComponent
Human Hiv-1 Protease (from E. coli)
Human Hiv-1 Protease (from E. coli)
Catalog # 101189-714
Supplier:  BioVendor
CAS Number:  
Human Hiv-1 Protease (from E. coli)
Catalog # 101189-714
Supplier:  BioVendor
Supplier Number:  RH1P0001
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Source:
    E. coli
  • Species:
    Human
  • Size:
    100 µg
  • Enzyme Name:
    Hiv-1 Protease
  • Purity:
    >95%
  • Sequence:
    PQITLWQRPL VTIKIGGQLK EALLDTGADD TVLEEMNLPG RWKPKMIGGI GGFIKVRQYD QILIEICGHK AIGTVLVGPT PVNIIGRNLL TQIGCTLNF
  • Cat. No.:
    101189-714
  • Supplier no.:
    RH1P0001

Specifications

About this item

Retroviral protease from HIV-1 virus is an enzyme important in the life cycle of the virus

Sequence: PQITLWQRPL VTIKIGGQLK EALLDTGADD TVLEEMNLPG RWKPKMIGGI GGFIKVRQYD QILIEICGHK AIGTVLVGPT PVNIIGRNLL TQIGCTLNF

It is expressed in the infected cells as a part of Gag-Pol polyprotein from which it is autocatalytycaly released after formation of immature viral particle. The enzyme subsequently cleaves the other parts of viral polyproteins causing the maturation of the virus. In HIV-infected patients the enzyme is a subject of intensive mutagenesis and mutants resistant to applied medcines are produced as a consequence of the seletion pressure. HIV-1 protease is active as a homodimer.

Ordering information: For research use only.