Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Source:E. coli
- Size:100 µg
- Protein Synonyms:HBx
- Protein/Peptide Name:Hepatitis B Virus Protein X Hepatitis B virus
- Sequence:MAARVCCQLD PARDVLCLRP VGAESRGRPV SGPFGTLPSP SSSAVPADHG AHLSLRGLPV CAFSSAGPCA LRFTSARRME TTVNAHQVLP KVLHKRTLGL SAMSTTDLEA YFKDCLFKDW EELGEEIRLK VFVLGGCRHK LVCSPAPCNF FTSA
- Cat. No.:101215-430
- Supplier no.:RD972038100
Hepatitis B virus X protein (HBx) is a 17 kD transcriptional coactivator that plays a significant role in the regulation of genes involved in inflammation and cell survival. It regulates many transcription factors including nuclear factor kappa B (NF-kappaB) and plays a key role in hepatocarcinogenesis. HBx facilitates the binding of cAMP response element binding protein (CREB) to its responsive element. HBx stabilizes the cellular coactivator ASC-2 through direct protein-protein interaction, affecting the regulation of genes actively transcribed in liver cancer cells.
Sequence: MAARVCCQLD PARDVLCLRP VGAESRGRPV SGPFGTLPSP SSSAVPADHG AHLSLRGLPV CAFSSAGPCA LRFTSARRME TTVNAHQVLP KVLHKRTLGL SAMSTTDLEA YFKDCLFKDW EELGEEIRLK VFVLGGCRHK LVCSPAPCNF FTSA
Other names: HBx