To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Size:100 µg
- Protein Synonyms:PCT
- Protein/Peptide Name:Procalcitonin
- Molecular Weight:12.7 kDa
- Sequence:MKHHHHHHAS APFRSALEGL PDPTALSEKE GRLLLAALVK AYVQRKNELE QEQEQETEGS SLDSSRAKRC SNLSTCVLGT YSKDLNNFHT FSGIGFGAET PGKKRDIASG LERGR
- Cat. No.:102830-710
- Supplier no.:RD472006100
Total 115 AA. MW: 12.7 kDa (calculated). UniProtKB acc.no. P41547 (Ala26-Arg130). N-terminal His-tag (10 extra AA). Protein identity confirmed by LC-MS/MS.
Sequence: MKHHHHHHAS APFRSALEGL PDPTALSEKE GRLLLAALVK AYVQRKNELE QEQEQETEGS SLDSSRAKRC SNLSTCVLGT YSKDLNNFHT FSGIGFGAET PGKKRDIASG LERGR
Other names: PCT