Order Entry
United States
ContactUsLinkComponent
 
Rat Secretagonin (from E. coli)
Catalog # 102552-958
Supplier:  BioVendor
CAS Number:  
undefined
Rat Secretagonin (from E. coli)
Catalog # 102552-958
Supplier:  BioVendor
Supplier Number:  RD372120100
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Source:
    E. coli
  • Species:
    Rat
  • Size:
    100 µg
  • Protein Synonyms:
    SECRET|SCGN
  • Protein/Peptide Name:
    Secretagonin
  • Molecular Weight:
    33.3 kDa
  • Sequence:
    MKHHHHHHAS MDNAHRQTQA HLDAACFWQI WQRFDKDEKG YIKETELDAF FDDLLAKFGI EDTLMEENVQ KMKEQLMVGH DISKEGRILM KELASMFLSE DENFLLFFRL ETPLDNSVEF MQIWRKYDAD SSGFISAAEL SNFLRDLFLH HKKVISEAEL EEYTSTMMKI FDRNKDGRLD LNDLARILAL QENFLLQFKM DASSTEERKR DFEKIFAHYD VSKTGALEGP EVDGFVKDMM ELVQPSISGV DLDKFREILL RHCDVNKDGK IQKSELALCL GLKINP
  • Cat. No.:
    102552-958
  • Supplier no.:
    RD372120100

Specifications

About this item

Total 286 AA. MW: 33.3 kDa (calculated). UniProtKB acc.no. Q6R556. N-terminal His-tag, 10extra AA

Sequence: MKHHHHHHAS MDNAHRQTQA HLDAACFWQI WQRFDKDEKG YIKETELDAF FDDLLAKFGI EDTLMEENVQ KMKEQLMVGH DISKEGRILM KELASMFLSE DENFLLFFRL ETPLDNSVEF MQIWRKYDAD SSGFISAAEL SNFLRDLFLH HKKVISEAEL EEYTSTMMKI FDRNKDGRLD LNDLARILAL QENFLLQFKM DASSTEERKR DFEKIFAHYD VSKTGALEGP EVDGFVKDMM ELVQPSISGV DLDKFREILL RHCDVNKDGK IQKSELALCL GLKINP

Other names: SCGN, SECRET

Ordering information: For research use only.