Order Entry
United States
ContactUsLinkComponent
Rat Recombinant Cystatin C (from E. coli)
Rat Recombinant Cystatin C (from E. coli)
Catalog # 200068-242
Supplier:  BioVendor
CAS Number:  
undefined
Rat Recombinant Cystatin C (from E. coli)
Catalog # 200068-242
Supplier:  BioVendor
Supplier Number:  RD372009100
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Rat
  • Size:
    100 µg
  • Protein Synonyms:
    Cystatin-3|Post G-globulin|Gamma-trace|Neuroendocrine basic polypeptide|Post-gamma-globulin|CST3
  • Protein/Peptide Name:
    Cystatin C
  • Molecular Weight:
    13.36 kDa
  • Sequence:
    MRGSHHHHHH GMASGTSRPP PRLLGAPQEA DASEEGVQRA LDFAVSEYNK GSNDAYHSRA IQVVRARKQL VAGINYYLDV EMGRTTCTKS QTNLTNCPFH DQPHLMRKAL CSFQIYSVPW KGTHTLTKSS CKNA
  • Cat. No.:
    200068-242
  • Supplier no.:
    RD372009100

Specifications

About this item

Cystatin C is a non-glycosilated basic single-chain protein consisting of 120 amino acids with a molecular weight of 13.36 kDa and is characterized by two disulfide bonds in the carboxy-terminal region. It belongs to the cystatins superfamilly which inactivates lysosomal cysteine proteinases, e.g. cathepsin B, H,.K, L and S. Imbalance between Cystatin C and cysteine proteinases is associated with inflammation, renal failure, cancer, Alzheimer's disease, multiple sclerosis and hereditary Cystatin C amyloid angiopathy. Its increased level has been found in patients with autoimune diseases, with colorectal tumors and in patients on dyalisis. Serum Cystatin C seems to be better marker of glomerular filtration rate than creatinine. On the other hand, low concentration of Cystatin C presents a risk factor for secondary cardiovascular events.

Sequence: MRGSHHHHHH GMASGTSRPP PRLLGAPQEA DASEEGVQRA LDFAVSEYNK GSNDAYHSRA IQVVRARKQL VAGINYYLDV EMGRTTCTKS QTNLTNCPFH DQPHLMRKAL CSFQIYSVPW KGTHTLTKSS CKNA

Other names: Post G-globulin, Cystatin-3, Neuroendocrine basic polypeptide, Gamma-trace, Post-gamma-globulin, CST3

Ordering information: For research use only.