Order Entry
United States
ContactUsLinkComponent
Human Recombinant LECT-2 (from E. coli)
Human Recombinant LECT-2 (from E. coli)
Catalog # 102830-680
Supplier:  BioVendor
CAS Number:  
undefined
Human Recombinant LECT-2 (from E. coli)
Catalog # 102830-680
Supplier:  BioVendor
Supplier Number:  RD172370100
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    100 µg
  • Protein Synonyms:
    LECT2|hLECT2|Leukocyte Cell-Derived Che
  • Protein/Peptide Name:
    LECT-2
  • Sequence:
    MKHHHHHHAS GPWANICAGK SSNEIRTCDR HGCGQYSAQR SQRPHQGVDI LCSAGSTVYA PFTGMIVGQE KPYQNKNAIN NGVRISGRGF CVKMFYIKPI KYKGPIKKGE KLGTLLPLQK VYPGIQSHVH IENCDSSDPT AYL
  • Cat. No.:
    102830-680
  • Supplier no.:
    RD172370100

Specifications

About this item

LECT2 acts as a chemotactic factor to neutrophils. It stimulates the proliferation of chondrocytes and osteoblasts. It is strongly expressed in liver and weakly in testis.

Sequence: MKHHHHHHAS GPWANICAGK SSNEIRTCDR HGCGQYSAQR SQRPHQGVDI LCSAGSTVYA PFTGMIVGQE KPYQNKNAIN NGVRISGRGF CVKMFYIKPI KYKGPIKKGE KLGTLLPLQK VYPGIQSHVH IENCDSSDPT AYL

Other names: LECT-2, hLECT2, LECT2

Ordering information: For research use only.