Order Entry
United States
ContactUsLinkComponent
Human Recombinant Cornifin-B (from E. coli)
Human Recombinant Cornifin-B (from E. coli)
Catalog # 101189-690
Supplier:  BioVendor
CAS Number:  
undefined
Human Recombinant Cornifin-B (from E. coli)
Catalog # 101189-690
Supplier:  BioVendor
Supplier Number:  RD172320100
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    100 µg
  • Protein Synonyms:
    SPR-IB|Small proline-rich protein IB|SPRR1B|14.9 kDa pancornulin
  • Protein/Peptide Name:
    Cornifin-B
  • Purity:
    >95%
  • Sequence:
    MKHHHHHHAS SQ­QQKQPCTP PPQLQQQQVK QPCQPPPQEP CIPKTKEPCH PKVPEPCHPK VPEPCQPKVP EPCHPKVPEP CPSIVTPAPA QQKTKQK
  • Cat. No.:
    101189-690
  • Supplier no.:
    RD172320100

Specifications

About this item

Cornifin-B occurs in keratinocytes. It is cross-linked to membrane proteins by transglutaminase which leads to formation of an insoluble envelope. It is expressed during differentiation in squamous tissues such as epidermis, esophagus, tongue or trachea.

Sequence: MKHHHHHHAS SQ­QQKQPCTP PPQLQQQQVK QPCQPPPQEP CIPKTKEPCH PKVPEPCHPK VPEPCQPKVP EPCHPKVPEP CPSIVTPAPA QQKTKQK

Other names: SPRR1B, 14.9 kDa pancornulin, Small proline-rich protein IB, SPR-IB

Ordering information: For research use only.